Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLAIN1 Rabbit anti-Human, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of SLAIN1.
Manufacturer: Abnova Corporation PAB25708
Specifications
SLAIN1 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of SLAIN1. | |
In PBS (0.05% sodium azide) | |
Liquid | |
SLAIN1 | |
A synthetic peptide corresponding to 16 amino acids at internal region of human SLAIN1. | |
Peptide affinity purification | |
RUO | |
Primary | |
122060 | |
Human |
ELISA, Immunohistochemistry | |
Unconjugated | |
Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
SLAIN1 | |
C13orf32|FLJ30046|MGC131899 | |
Rabbit | |
IgG | |
100 ug | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
At least three isoforms of SLAIN1 are known to exist; this antibody will detect only the largest isoform. SLAIN1 antibody is predicted not to cross-react with SLAIN2. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only