Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLAIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of SLAIN1.

Manufacturer:  Abnova Corporation PAB25786

Catalog No. 89-127-412


Add to cart

Description

Description

Specifications

Specifications

SLAIN1
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of SLAIN1.
In PBS (0.02% sodium azide)
Liquid
SLAIN1
A synthetic peptide corresponding to 15 amino acids at C-terminus of human SLAIN1.
Peptide affinity purification
RUO
Primary
122060
Human, Murine, Rat
ELISA, Immunofluorescence, Western Blot
Unconjugated
Western Blot (1 ug/mL) Immunofluorescence (2.5 ug/mL) The optimal working dilution should be determined by the end user.
SLAIN1
C13orf32|FLJ30046|MGC131899
Rabbit
IgG
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
At least three isoforms of SLAIN1 are known to exist; this antibody will detect all three. SLAIN1 antibody is predicted not to cross-react with SLAIN2.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only