Learn More
Slc12a1, Rabbit, HRP, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.
Supplier:  Abnova Corporation PAB29049
Catalog No. 89-130-562
Add to Cart
Description
Description
Specifications
Specifications
Slc12a1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
MGC93479/Nkcc2
Rabbit
Affinity purification
RUO
25065
Human, Mouse, Rat
Liquid
Western Blot
HRP
Western blot (1:1000) The optimal working dilution should be determined by the end user.
Slc12a1
Slc12a1
A synthetic peptide corresponding to amino acids 33-55 at N-terminus of rat Slc12a1.
100 μg
Primary
Detects ∼160 kDa.
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions