Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC16A12, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of SLC16A12.

Manufacturer:  Abnova Corporation PAB17772

Catalog No. 89-119-438


Add to cart

Description

Description

Specifications

Specifications

SLC16A12
Unconjugated
Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user.
SLC16A12
CJMG|DKFZp686E188|MCT12
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects endogenous levels of total SLC16A12 protein.
ELISA, Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of SLC16A12.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Liquid
SLC16A12
A synthetic peptide corresponding to internal of human SLC16A12.
RUO
Primary
387700
Human, Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only