Learn More
SLC2A9 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Slc2a9.
Manufacturer:  Abnova Corporation PAB12029
Catalog No. 89-115-538
Add to cart
Description
Description
Specifications
Specifications
SLC2A9
Unconjugated
Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Slc2a9
Glut9
Rabbit
100 uL
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Mouse
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Slc2a9.
In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Liquid
Slc2a9
A synthetic peptide corresponding to amino acids 400-450 of mouse Slc2a9.
RUO
Primary
117591