Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC4A11 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of SLC4A11.

Manufacturer:  Abnova Corporation PAB16074

Catalog No. 89-118-041


Add to cart

Description

Description

Specifications

Specifications

SLC4A11
Unconjugated
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
SLC4A11
BTR1|CDPD|CDPD1|CHED2|MGC126418|MGC126419|NABC1|dJ794I6.2
Rabbit
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of SLC4A11.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Liquid
SLC4A11
A synthetic peptide corresponding to C-terminus of human SLC4A11.
RUO
Primary
83959
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only