Learn More
SLIRP, Rabbit, HRP, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLIRP.
Manufacturer:  Abnova Corporation PAB15210
Catalog No. 89-117-505
Add to cart
Description
Description
Steroid receptor RNA activator (SRA, or SRA1; MIM 603819) is a complex RNA molecule containing multiple stable stem-loop structures that functions in coactivation of nuclear receptors. SLIRP interacts with stem-loop structure-7 of SRA (STR7) and modulates nuclear receptor transactivation (Hatchell et al., 2006 [PubMed 16762838]).[supplied by OMIM
Specifications
Specifications
SLIRP
HRP
Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
SLIRP
DC23|C14orf156|DC50|PD04872
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
Does not react with mouse. This is specific to SLIRP protein. This antibody may be used for Western blot analysis where a band at ~12 KDa is be seen, representing the SLIRP protein. May see additional bands above 45 KDa.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of SLIRP.
In PBS
Liquid
SLIRP
A synthetic peptide corresponding to C-terminus of human SLIRP.
RUO
Primary
81892
Human, Primate