Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLO, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant SLO.

Manufacturer:  Abnova Corporation PAB15598

Catalog No. 89-138-111

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

SLO
Unconjugated
Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to hemolytic Streptococcus SLO.
RUO
Primary
This antibody react with Streptolysin O (Streptolysin O of Group A, C, and G origins).
Inhibition Assays, Western Blot
Rabbit polyclonal antibody raised against recombinant SLO.
In antiserum (0.09% sodium azide)
Rabbit
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Streptococcus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only