Learn More
SMAD1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SMAD1.
Supplier: Abnova Corporation PAB26822
Description
The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq
Sequence: I-S-S-V-SpSpecifications
SMAD1 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
SMAD1 | |
SMAD1 | |
A synthetic peptide corresponding to residues surrounding S465 of human SMAD1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of SMAD1. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
BSP1/JV4-1/JV41/MADH1/MADR1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4086 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only