Learn More
SMAD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SMAD1.
Supplier:  Abnova Corporation PAB5579
Catalog No. 89-131-915
Add to Cart
Description
Description
The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq
Specifications
Specifications
SMAD1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SMAD1.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
BSP1/JV4-1/JV41/MADH1/MADR1
Rabbit
100 μg
Primary
Human
Liquid
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
SMAD1
SMAD1
A synthetic peptide corresponding to N-terminus of human SMAD1.
RUO
4086
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions