Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMO Rabbit anti-Bovine, Dog, Fruit fly, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SMO.
Supplier: Abnova Corporation PAB26572
Specifications
SMO | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SMO. | |
In PBS (0.09% sodium azide) | |
Gx/SMOH | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
6608 | |
Bovine, Canine, Drosophila, Equine, Hamster, Human, Monkey, Mouse, Porcine, Rabbit, Rat | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
SMO | |
SMO | |
A synthetic peptide corresponding to human SMO. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only