Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNCA Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant SNCA.

Supplier:  Abnova Corporation PAB5100

Catalog No. 89-131-744


Add to Cart

Description

Description

Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. Synucleins are abundantly expressed in the brain and alpha- and beta-synuclein inhibit phospholipase D2 selectively. SNCA may serve to integrate presynaptic signaling and membrane trafficking. Defects in SNCA have been implicated in the pathogenesis of Parkinson disease. SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Four alternatively spliced transcripts encoding two different isoforms have been identified for this gene. [provided by RefSeq

Specifications

Specifications

SNCA
Polyclonal
Rabbit polyclonal antibody raised against recombinant SNCA.
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
MGC110988/NACP/PARK1/PARK4/PD1
Rabbit
150 μg
Primary
Human
Liquid
ELISA, Immunoprecipitation
Unconjugated
The optimal working dilution should be determined by the end user.
SNCA
SNCA
Recombinant protein corresponding to human SNCA.
RUO
6622
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only