Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA6, Rabbit, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of SPATA6.
Manufacturer: Abnova Corporation PAB25743
Specifications
SPATA6 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of SPATA6. | |
In PBS (0.02% sodium azide) | |
Liquid | |
SPATA6 | |
A synthetic peptide corresponding to 14 amino acids at C-terminus of human SPATA6. | |
Peptide affinity purification | |
RUO | |
Primary | |
54558 | |
Human, Murine, Rat |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
SPATA6 | |
FLJ10007|SRF-1|SRF1 | |
Rabbit | |
IgG | |
100 ug | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
At least three isoforms of SPATA6 are known to exist; this antibody will detect only the longest two isoforms. SPATA6 antibody is predicted to not cross-react with other SPATA protein family members. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only