Learn More
SSTR2 Rabbit anti-Bovine, Chicken, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SSTR2.
Supplier:  Abnova Corporation PAB26524
Catalog No. 89-128-164
Add to Cart
Description
Description
Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The biologic effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner. SSTR2 is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in cerebrum and kidney. [provided by RefSeq
Specifications
Specifications
SSTR2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SSTR2.
In PBS (0.09% sodium azide)
SSTR2
A synthetic peptide corresponding to 16 amino acids at C-terminus of human SSTR2.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (16 ug/mL) The optimal working dilution should be determined by the end user.
SSTR2
Rabbit
Immunoaffinity chromatography
RUO
6752
Bovine, Canine, Chicken, Equine, Hamster, Human, Monkey, Mouse, Porcine, Rat
Liquid
Recommendations
Recommendations