Learn More
SSTR5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SSTR5.
Supplier:  Abnova Corporation PAB26474
Catalog No. 89-128-113
Add to Cart
Description
Description
Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The biological effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner. SSTR5 is a member of the superfamily of receptors having seven transmembrane segments. [provided by RefSeq
Specifications
Specifications
SSTR5
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SSTR5.
In PBS (0.09% sodium azide)
SSTR5
A synthetic peptide corresponding to 17 amino acids at C-terminus of human SSTR5.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (23 ug/mL) The optimal working dilution should be determined by the end user.
SSTR5
Rabbit
Immunoaffinity chromatography
RUO
6755
Human
Liquid
Product Suggestions
Product Suggestions