Learn More
STARD13 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STARD13.
Supplier: Abnova Corporation PAB17478
Description
This gene encodes a protein that contains a sterile alpha motif domain in the N-terminus, an ATP/GTP-binding motif, a GTPase-activating protein domain, and a STAR-related lipid transfer domain in the C-terminus. The gene is located in a region of chromosome 13 that has loss of heterozygosity in hepatic cancer. At least three alternatively spliced transcript variants have been described for this gene. [provided by RefSeq
Specifications
STARD13 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of STARD13. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
DLC2/FLJ37385/GT650 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total STARD13 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence | |
Unconjugated | |
Immunofluorescence (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
STARD13 | |
STARD13 | |
A synthetic peptide corresponding to internal of human STARD13. | |
RUO | |
90627 | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only