Learn More
STAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STAT3.
Supplier:  Abnova Corporation PAB27226
Catalog No. 89-128-881
Add to Cart
Description
Description
The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated through phosphorylation in response to various cytokines and growth factors including IFNs, EGF, IL5, IL6, HGF, LIF and BMP2. This protein mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of this protein. PIAS3 protein is a specific inhibitor of this protein. Three alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Specifications
Specifications
STAT3
Polyclonal
Unconjugated
Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user.
STAT3
STAT3
A synthetic peptide corresponding to 19 amino acids at N-terminus of human STAT3.
100 μg
Primary
Human, Mouse, Rat
Liquid
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of STAT3.
In PBS (0.02% sodium azide)
APRF/FLJ20882/HIES/MGC16063
Rabbit
Peptide affinity purification
RUO
6774
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions
Product Suggestions