Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-STAT5a (Methyl Arg31), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244184

Catalog No. NBP244184


Add to cart

Description

Description

Specifically detects STAT5a (Methyl Arg31) in Human samples, and it is validated for ELISA, Western Blotting

STAT5A Methyl Arg31 Polyclonal antibody specifically detects STAT5A Methyl Arg31 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

STAT5a (Methyl Arg31)
0.57 mg/mL
Western Blot 1:500, ELISA 1:5000
Affinity Purified
STAT5A
STAT5A [Arg31-Methyl] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic monomethylated peptide surrounding Arg31 of human STAT.
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with 0.05% Sodium Azide
MGF, signal transducer and activator of transcription 5A, STAT5
Rabbit
IgG
0.1 mg
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary
6776
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only