Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STMN, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of STMN.

Manufacturer:  Abnova Corporation PAB14834

Catalog No. 89-138-019

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

STMN
Lot specific
Rabbit polyclonal antibody raised against synthetic peptide of STMN.
In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Rabbit
Immunoaffinity purification
RUO
Primary
Epitope: Internal region.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to internal region of human STMN.
100 uL
Store at 4°C. Do not freeze.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only