Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STRN4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP236537
Description
STRN4 Polyclonal specifically detects STRN4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
STRN4 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
FLJ35594, striatin, calmodulin binding protein 4, striatin-4, ZIN, zinedin | |
Rabbit | |
Affinity Purified | |
RUO | |
29888 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
NP_037535.2 | |
STRN4 | |
The immunogen recognized by the antibody maps to a region between residue 325 to 375 of human Striatin-4 using the numbering given in entry NP_037535.2. | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction