Learn More
SUCNR1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SUCNR1.
Supplier:  Abnova Corporation PAB26400
Catalog No. 89-128-041
Add to Cart
Description
Description
Specifications
Specifications
SUCNR1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SUCNR1.
In PBS (0.09% sodium azide)
GPR91
Rabbit
Immunoaffinity chromatography
RUO
56670
Human, Monkey
Liquid
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3 ug/mL) The optimal working dilution should be determined by the end user.
SUCNR1
SUCNR1
A synthetic peptide corresponding to 15 amino acids at 2nd cytoplasmic domain of human SUCNR1.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations