Learn More
Swine H1N1 Hemagglutinin Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Swine H1N1 Hemagglutinin.
Manufacturer:  Abnova Corporation PAB16777
Catalog No. 89-138-229
Add to cart
Description
Description
Specifications
Specifications
Swine H1N1 Hemagglutinin
Unconjugated
ELISA (2 ng of free peptide at 1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to Swine H1N1 Hemagglutinin.
RUO
Primary
The peptide sequence is unique from the peptide sequence for product PAB16773 and PAB16769. This antibody is a cognate pair with product number PAB16775.
ELISA
Rabbit polyclonal antibody raised against synthetic peptide of Swine H1N1 Hemagglutinin.
In PBS (0.02% sodium azide)
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Virus