Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Swine Serum, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244106

Catalog No. NBP244106


Add to cart

Description

Description

Specifically detects Swine Serum in Porcine samples, and it is validated for ELISA, Immunoprecipitation, Western Blotting

Swine Serum Polyclonal antibody specifically detects Swine Serum in Porcine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications

Specifications

Swine Serum
LYOPH
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Delipidation and Defibrination
Porcine Serum Proteins.
Delipidation and Defibrination
RUO
Primary
Reconstitute with 2.0 mL deionized water.
Western Blot, ELISA, Immunoprecipitation
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Rabbit
Serum
2 ml
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only