Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYCP1 Rabbit anti-Mouse, DyLight 649, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Sycp1.
Supplier: Abnova Corporation PAB14963
Specifications
SYCP1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Sycp1. | |
In 50mM sodium borate | |
SCP1 | |
Rabbit | |
100 μL | |
Primary | |
This is specific to mouse SCP1. This antibody is useful for immunofluorescence and immunohistochemistry (paraffin and frozen sections). | |
Store at 4°C. Do not freeze. |
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed) | |
DyLight 649 | |
Immunofluorescence (1:100-1:750) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:750) Immunohistochemistry (Frozen sections) (1:750) The optimal working dilution should be determined by the end user. | |
Sycp1 | |
Sycp1 | |
A synthetic peptide corresponding to C-terminus of mouse Sycp1. | |
RUO | |
20957 | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction