Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYCP3 Rabbit anti-Human, Mouse, HRP, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SYCP3.
Supplier: Abnova Corporation PAB15236
Specifications
SYCP3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SYCP3. | |
In PBS | |
COR1/MGC71888/SCP3 | |
Rabbit | |
100 μL | |
Primary | |
This SCP3 antibody recognizes human and murine protein. Other species have not been tested. | |
Store at 4°C. Do not freeze. |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
HRP | |
Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
SYCP3 | |
SYCP3 | |
A synthetic peptide corresponding to C-terminus of human SYCP3. | |
RUO | |
50511 | |
Human, Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction