Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYF2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SYF2
Supplier: Abnova Corporation PAB28017
Description
This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: AAIAASEVLVDSAEEGSLAAAAELAAQKREQRLRKFRELHLMRNEARKLNHQEVVEEDKRLKLPANWEAKKARLEWELKEEEKKKECAARGSpecifications
SYF2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant SYF2 | |
In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
CBPIN/DKFZp564O2082/NTC31/P29/fSAP29 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
25949 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:500 - 1:1000) Western Blot (1:100 - 1:250) The optimal working dilution should be determined by the end user. | |
SYF2 | |
SYF2 | |
Recombinant protein corresponding to amino acids of human SYF2 | |
100 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only