Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYK Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of SYK.

Manufacturer:  Abnova Corporation PAB18225

Catalog No. 89-119-795


Add to cart

Description

Description

Specifications

Specifications

SYK
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of SYK.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Liquid
SYK
A synthetic peptide corresponding to human SYK.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody is specific to SYK.
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user.
SYK
DKFZp313N1010|FLJ25043|FLJ37489
Rabbit
Affinity purification
RUO
Primary
6850
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only