Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T7 tag Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against T7 tag.

Manufacturer:  Abnova Corporation PAB0884

Catalog No. 89-136-904


Add to cart

Description

Description

Sequence: MASMTGGQQMG
Specifications

Specifications

T7 tag
Unconjugated
Western Blot (1:400) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to T7 tag.
RUO
Primary
This antibody recognizes proteins that contain the T7 epitope tag.
Western Blot
Rabbit polyclonal antibody raised against T7 tag.
In PBS (0.2% gelatin, 0.09% sodium azide)
Rabbit
200 ug
Store at 4°C. Do not freeze.
Polyclonal
Not Provided
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only