Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF2 Rabbit anti-Yeast, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of TAF2.

Manufacturer:  Abnova Corporation PAB15971

Catalog No. 89-117-990


Add to cart

Description

Description

Specifications

Specifications

TAF2
Unconjugated
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
TAF2
TAF150|TSM1
Rabbit
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Yeast
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of TAF2.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Liquid
TAF2
A synthetic peptide corresponding to N-terminus of Saccharomyces cerevisiae TAF2.
RUO
Primary
850409
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only