Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAOK1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Taok1.
Manufacturer: Abnova Corporation PAB17946
Specifications
TAOK1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Taok1. | |
In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
Taok1 | |
A synthetic peptide corresponding to the residues surrounding amino acid 414 of rat Taok1. | |
RUO | |
286993 | |
Human, Mouse, Rat |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
Taok1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼63 KDa bands corresponding to TAOK1 in samples from human origin. | |
Liquid |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only