Learn More
TAS1R1 Rabbit anti-Bovine, Dog, Horse, Human, Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TAS1R1.
Supplier: Abnova Corporation PAB27727
Description
The protein encoded by this gene is a G protein-coupled receptor and is a component of the heterodimeric amino acid taste receptor T1R1+3. The T1R1+3 receptor responds to L-amino acids but not to D-enantiomers or other compounds. Most amino acids that are perceived as sweet activate T1R1+3, and this activation is strictly dependent on an intact T1R1+3 heterodimer. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq
Specifications
TAS1R1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TAS1R1. | |
In PBS (0.09% sodium azide) | |
GPR70/T1R1/TR1/gm148 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
80835 | |
Bovine, Canine, Equine, Human, Porcine | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6 ug/mL) The optimal working dilution should be determined by the end user. | |
TAS1R1 | |
TAS1R1 | |
A synthetic peptide corresponding to 16 amino acid at cytoplasmic domain of human TAS1R1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins, except TAS1R2 (69%). | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only