Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAS1R2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TAS1R2.
Supplier: Abnova Corporation PAB27728
Specifications
TAS1R2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TAS1R2. | |
In PBS (0.09% sodium azide) | |
GPR71/T1R2/TR2 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
80834 | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-9 ug/mL) The optimal working dilution should be determined by the end user. | |
TAS1R2 | |
TAS1R2 | |
A synthetic peptide corresponding to 20 amino acid at N-terminus of human TAS1R2. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only