Learn More
TAS1R3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TAS1R3.
Manufacturer:  Abnova Corporation PAB25123
Catalog No. 89-126-797
Add to cart
Description
Description
The TAS1R3 gene encodes the human homolog of mouse Sac, a major determinant of differences between sweet-sensitive and -insensitive mouse strains in their responsiveness to sucrose, saccharine, and other sweeteners.[supplied by OMIM
Specifications
Specifications
TAS1R3
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of TAS1R3.
In PBS, pH 7.2 (0.05% sodium azide)
Liquid
TAS1R3
A synthetic peptide corresponding to TAS1R3.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
TAS1R3 polyclonal antibody detects endogenous levels of TAS1R3 protein.
Immunofluorescence, Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
TAS1R3
T1R3
Rabbit
Affinity purification
RUO
Primary
83756
Human