Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D10A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP236551
Description
TBC1D10A Polyclonal specifically detects TBC1D10A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
TBC1D10A | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
EBP50-PDX interactor of 64 kDa, EPI64, rab27A-GAP-alpha, TBC1 domain family member 10A, TBC1 domain family, member 10A | |
Rabbit | |
Affinity Purified | |
RUO | |
83874 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
NP_114143.1 | |
TBC1D10A | |
The immunogen recognized by the antibody maps to a region between residue 458 to 508 of human EBP50-PDX Interactor of 64 kDa using the numbering given in entry NP_114143.1. | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction