Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D30 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant TBC1D30.

Supplier:  Abnova Corporation PAB27509

Catalog No. 89-129-018


Add to Cart

Description

Description

Sequence: NSSPVINHLLLGKKMKMTNRAAKNAVIHIPGHTGGKISPVPYEDLKTKLNSPWRTHIRVHKKNMPRTKSHPGCGDTVGLIDEQNEASKTNGLG
Specifications

Specifications

TBC1D30
Polyclonal
Rabbit polyclonal antibody raised against recombinant TBC1D30.
In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide)
KIAA0984
Rabbit
Antigen affinity purification
RUO
23329
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
TBC1D30
TBC1D30
Recombinant protein corresponding to amino acids of human TBC1D30.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only