Learn More
Rabbit anti-TEF1, Clone: 13112, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243581
Catalog No. NBP243581
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunoprecipitation
TEF1 Monoclonal antibody specifically detects TEF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
TEF1
13112
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500
Purified
TEAD1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human TEF1. The exact sequence is proprietary.
48 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
PBS with No Preservative
NTEF-1, protein GT-IIC, TEA domain family member 1 (SV40 transcriptional enhancer factor), TEAD-1
Mouse
IgG
Protein G purified
RUO
Primary
7003