Learn More
TFEC Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP243766
Catalog No. NBP243766
Add to Cart
Description
Description
Specifically detects TFEC in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
TFEC Polyclonal specifically detects TFEC in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
TFEC
Polyclonal
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Class E basic helix-loop-helix protein 34, hTFEC-L, TCFECTFECLbHLHe34BHLHE34, TFE-C, transcription factor EC, Transcription factor EC-like
Rabbit
39 kDa
0.1 mL
Primary
Human
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
TFEC
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human TFEC. The exact sequence is proprietary.
Immunogen affinity purified
RUO
22797
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions
Product Suggestions