Learn More
TICAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of TICAM2.
Manufacturer:  Abnova Corporation PAB19153
Catalog No. 89-120-679
Add to cart
Description
Description
TIRP is a Toll/interleukin-1 receptor (IL1R; MIM 147810) (TIR) domain-containing adaptor protein involved in Toll receptor signaling (see TLR4; MIM 603030).[supplied by OMIM
Specifications
Specifications
TICAM2
1 mg/mL
Rabbit polyclonal antibody rasied against synthetic peptide of TICAM2.
In PBS (0.02% sodium azide)
Liquid
TICAM2
A synthetic peptide corresponding near C-terminus of human TICAM2.
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
ELISA, Immunohistochemistry, Western Blot
Unconjugated
Western blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
TICAM2
MGC129876|MGC129877|TICAM-2|TIRAP3|TIRP|TRAM
Rabbit
Ion exchange chromatography purification
RUO
Primary
353376