Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR8 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Tlr8.
Manufacturer: Abnova Corporation PAB17930
Specifications
TLR8 | |
Polyclonal | |
toll-like receptor 8 | |
Tlr8 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∽120kDa TLR8 in samples from human, mouse and rat origins. | |
Liquid |
Western Blot | |
Unconjugated | |
In PBS pH 7.2 (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
Tlr8 | |
A synthetic peptide corresponding to the residues surrounding amino acid 942 of mouse Tlr8. | |
RUO | |
170744 | |
Human, Mouse, Rat |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only