Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant TNF.

Manufacturer:  Abnova Corporation PAB16156

Catalog No. 89-118-055


Add to cart

Description

Description

This gene encodes a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophages. It can bind to, and thus functions through its receptors TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer. Knockout studies in mice also suggested the neuroprotective function of this cytokine. [provided by RefSeq

Specifications

Specifications

TNF
Unconjugated
ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
TNF
DIF|TNF-alpha|TNFA|TNFSF2
Rabbit
IgG
RUO
Primary
7124
ELISA
Rabbit polyclonal antibody raised against recombinant TNF.
Lyophilized from PBS.
Lyophilized
TNF
Recombinant protein corresponding to human TNF.
100 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only