Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF10A Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89134242 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-242 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-242 Supplier Abnova Corporation Supplier No. PAB9573

Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF10A.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10/TRAIL), and thus transduces cell death signal and induces cell apoptosis. Studies with FADD-deficient mice suggested that FADD, a death domain containing adaptor protein, is required for the apoptosis mediated by this protein. [provided by RefSeq

Specifications

Antigen TNFRSF10A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description tumor necrosis factor receptor superfamily, member 10a
Formulation In PBS, pH 7.2 (30% glycerol, 0.5mg/mL BSA, 0.01% thimerosal)
Gene TNFRSF10A
Gene Alias APO2, CD261, DR4, MGC9365, TRAILR-1, TRAILR1
Gene Symbols TNFRSF10A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding amino acids 454 of human TNFRSF10A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8797
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.