Learn More
TNFSF8, Rabbit, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of TNFSF8.
Manufacturer: Abnova Corporation PAB26949
Description
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF8/CD30, which is a cell surface antigen and a marker for Hodgkin lymphoma and related hematologic malignancies. The engagement of this cytokine expressed on B cell surface plays an inhibitory role in modulating Ig class switch. This cytokine was shown to enhance cell proliferation of some lymphoma cell lines, while to induce cell death and reduce cell proliferation of other lymphoma cell lines. The pleiotropic biologic activities of this cytokine on different CD30+ lymphoma cell lines may play a pathophysiologic role in Hodgkin's and some non-Hodgkin's lymphomas. [provided by RefSeq
Specifications
TNFSF8 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of TNFSF8. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
Liquid | |
TNFSF8 | |
A synthetic peptide corresponding to human TNFSF8. | |
100 uL | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
TNFSF8 polyclonal antibody detects endogenous levels of TNFSF8 protein. |
Immunofluorescence, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
TNFSF8 | |
CD153|CD30L|CD30LG|MGC138144 | |
Rabbit | |
Affinity purification | |
RUO | |
Primary | |
944 | |
Human, Murine |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only