Learn More
TP53 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TP53.
Supplier: Abnova Corporation PAB26665
Description
This gene encodes tumor protein p53, which responds to diverse cellular stresses to regulate target genes that induce cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism. p53 protein is expressed at low level in normal cells and at a high level in a variety of transformed cell lines, where it's believed to contribute to transformation and malignancy. p53 is a DNA-binding protein containing transcription activation, DNA-binding, and oligomerization domains. It is postulated to bind to a p53-binding site and activate expression of downstream genes that inhibit growth and/or invasion, and thus function as a tumor suppressor. Mutants of p53 that frequently occur in a number of different human cancers fail to bind the consensus DNA binding site, and hence cause the loss of tumor suppressor activity. Alterations of this gene occur not only as somatic mutations in human malignancies, but also as germline mutations in some cancer-prone families with Li-Fraumeni syndrome. Multiple p53 variants due to alternative promoters and multiple alternative splicing have been found. These variants encode distinct isoforms, which can regulate p53 transcriptional activity. [provided by RefSeq]
Sequence: P-Q-Sp-D-PSpecifications
TP53 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
TP53 | |
TP53 | |
A synthetic peptide corresponding to residues surrounding S6 of human TP53. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of TP53. | |
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
FLJ92943/LFS1/TRP53/p53 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7157 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only