Learn More
TP53 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TP53.
Manufacturer:  Abnova Corporation PAB14117
Catalog No. 89-137-831
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
TP53
Unconjugated
Western Blot (1:200) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to amino acids surrounding methylated K372 of TP53.
RUO
Primary
Human
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of TP53.
In PBS (0.05% sodium azide, 0.05% proclin 300)
Rabbit
50 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal