Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP53INP1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of TP53INP1.

Manufacturer:  Abnova Corporation PAB13806

Catalog No. 89-116-849


Add to cart

Description

Description

Specifications

Specifications

TP53INP1
Unconjugated
Western Blot (1-2 ug/mL) Immunohistochemistry (2 ug/mL) The optimal working dilution should be determined by the end user.
TP53INP1
Liquid
TP53INP1
A synthetic peptide corresponding to N-terminus 14 amino acids of human TP53INP1.
RUO
Primary
94241
Human, Murine, Rat
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of TP53INP1.
In PBS (0.02% sodium azide)
Q96A56
DKFZp434M1317|FLJ22139|SIP|TP53DINP1|TP53INP1A|TP53INP1B|Teap|p53DINP1
Rabbit
100 ug
Store at 4°C.
Polyclonal
Recognizes p53DINP1.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only