Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM25 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of TRIM25.

Manufacturer:  Abnova Corporation PAB13293

Catalog No. 89-116-542


Add to cart

Description

Description

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. The presence of potential DNA-binding and dimerization-transactivation domains suggests that this protein may act as a transcription factor, similar to several other members of the TRIM family. Expression of the gene is upregulated in response to estrogen, and it is thought to mediate estrogen actions in breast cancer as a primary response gene. [provided by RefSeq

Specifications

Specifications

TRIM25
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
TRIM25
EFP|RNF147|Z147|ZNF147
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine
Immunocytochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of TRIM25.
In PBS (0.02% sodium azide)
Liquid
TRIM25
A synthetic peptide corresponding to C-terminus 19 amino acids of human TRIM25.
RUO
Primary
7706
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only