Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trpc6 Rabbit anti-Mouse, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Trpc6.

Manufacturer:  Abnova Corporation PAB14886

Catalog No. 89-117-331


Add to cart

Description

Description

Specifications

Specifications

Trpc6
Unconjugated
ELISA (1 ug/mL) Immunohistochemistry (5-10 ug/mL) Immunocytochemistry (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Trpc6
AV025995|LLHWJM002|LLHWJM003|LLHWJM004|Trrp6|mtrp6
Rabbit
25 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Murine, Murine, Rabbit, Rabbit
ELISA, Immunocytochemistry, Immunohistochemistry
Rabbit polyclonal antibody raised against synthetic peptide of Trpc6.
In PBS (0.1% proclin, 2% Block Ace)
Liquid
Trpc6
A synthetic peptide corresponding to C-terminus of mouse Trpc6.
RUO
Primary
22068
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only