Learn More
Trpm2 Rabbit anti-Mouse, Rat, Biotin, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Trpm2.
Manufacturer:  Abnova Corporation PAB15111
Catalog No. 89-117-453
Add to cart
Description
Description
Specifications
Specifications
Trpm2
Biotin
Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Trpm2
9830168K16Rik|C79133|LTRPC2|TRPC7|Trp7|Trrp7
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody is useful in Western blot, where a band is seen ~172 KDa.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Trpm2.
In PBS
Liquid
Trpm2
A synthetic peptide corresponding to amino acids 1200-1300 of mouse Trpm2.
RUO
Primary
28240
Murine, Rat