Learn More
TRPV2 Rabbit anti-Horse, Human, Monkey, Mouse, Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRPV2.
Manufacturer:  Abnova Corporation PAB16478
Catalog No. 89-118-351
Add to cart
Description
Description
This gene encodes an ion channel that is activated by high temperatures above 52 degrees Celsius. The protein may be involved in transduction of high-temperature heat responses in sensory ganglia. It is thought that in other tissues the channel may be activated by stimuli other than heat. [provided by RefSeq
Specifications
Specifications
TRPV2
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
TRPV2
MGC12549|VRL|VRL-1|VRL1
Rabbit
Immunoaffinity purification
RUO
Primary
51393
Equine, Human, Monkey, Murine, Porcine
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of TRPV2.
In PBS (0.09% sodium azide)
Liquid
TRPV2
A synthetic peptide (conjugated with KLH) corresponding to human TRPV2.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
N-terminal domain of human.