Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trypsin Inhibitor Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244238
Description
Specifically detects Trypsin Inhibitor in C. elegans, Plant samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting
Trypsin Inhibitor Polyclonal specifically detects Trypsin Inhibitor in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Trypsin Inhibitor | |
Polyclonal | |
HRP | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate | |
KTI3, Kunitz trypsin inhibitor | |
Rabbit | |
Multi-step | |
RUO | |
547831 | |
Plant | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
LYOPH | |
Western Blot 1:1000 - 1:5000, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000 | |
P01070 | |
KTI3 | |
Trypsin Inhibitor [Lima Bean] (Uniprot: P01070) | |
0.1 mg | |
Primary | |
Reconstitute with 100 ul deionized water (or equivalent) | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction